Learn More
Cayman Chemical AvenanthramideC methyl ester
SDP
Supplier:  Cayman Chemical 10011336500 UG
Encompass_Preferred
Inhibitor of NF-κB activation that blocks the phosphorylation of IKK and IκB (IC50 ~ 40 μM); dose dependently inhibits the expression and secretion of IL-6, IL-8, and MCP-1 in human aortic endothelial cells.
Catalog No. 50-288-9543