Learn More
AdipoGen Averantin
SDP
Supplier:  AdipoGen BVT0169M001
Encompass_Preferred
Chemical. CAS 5803-62-3. Formula C20H20O7. MW 372.4. Isolated from Aspergillus sp. strain WDMH51. Mycotoxin. Intermediate of the biosynthetic pathway to aflatoxin B1. Cytotoxic against human solid tumor cell lines.
Catalog No. 50-256-2080