Learn More
Abcam Averantin, Aflato x in B1 percursor, 5MG
Catalog No. 502705996
Encompass_Preferred
MW 372.4 Da, Purity >98%. Mycotoxin. Aflatoxin B1 precursor. Cytotoxic against solid tumor cell lines.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5996 Supplier Abcam Supplier No. AB1442015MG
MW 372.4 Da, Purity >98%. Mycotoxin. Aflatoxin B1 precursor. Cytotoxic against solid tumor cell lines.
The product is subject to the following: Abcam Restricted Use Statement