Learn More
Cayman Chemical AvermectIn B1a 1mg
SDP
Catalog No. 502987079
Encompass_Preferred
An anthelmintic and insecticidal agent; binds to high and low affinity sites on the mammalian GABAA receptor to activate or block calcium influx, respectively; inhibits binding of strychnine to the glycine receptor (Kis = 1.3 and 3.6 µM, respectively)
Catalog No. 50-298-7079 Supplier Cayman Chemical Supplier No. 220001 MG
An anthelmintic and insecticidal agent; binds to high and low affinity sites on the mammalian GABAA receptor to activate or block calcium influx, respectively; inhibits binding of strychnine to the glycine receptor (Kis = 1.3 and 3.6 µM, respectively)