Learn More
Medchemexpress LLC Avermectin B1a | 65195-55-3 | C48H72O14 | 100 MG
SDP
Catalog No. 5000373276
Encompass_Preferred
Avermectin B1a is an antiparasitic agent that paralyzes nematodes without causing hypercontraction or flaccid paralysis. It preferentially binds to synaptic membranes from several regions of rat brain. Its specific binding to intact monolayers of granule cells increases rapidly with time of incubation and reaches equilibrium after approximately 20 minutes at 24°C.
  • Antiparasitic agent
  • Paralyzes nematodes without causing hypercontraction or flaccid paralysis
  • Binds to synaptic membranes in rat brain
  • Activates GABA-insensitive chloride channels
  • Potency similar to moxidectin (IC50 values, 120 and 126 nM, respectively)
Catalog No. 50-003-73276 Supplier Medchemexpress LLC Supplier No. HY15308100MG
Avermectin B1a is an antiparasitic agent that paralyzes nematodes without causing hypercontraction or flaccid paralysis. It preferentially binds to synaptic membranes from several regions of rat brain. Its specific binding to intact monolayers of granule cells increases rapidly with time of incubation and reaches equilibrium after approximately 20 minutes at 24°C.
  • Antiparasitic agent
  • Paralyzes nematodes without causing hypercontraction or flaccid paralysis
  • Binds to synaptic membranes in rat brain
  • Activates GABA-insensitive chloride channels
  • Potency similar to moxidectin (IC50 values, 120 and 126 nM, respectively)