Learn More
Cayman Chemical Avermectin B1a aglycone, Purity: ≥95%, CAS: 71828-14-3, 1 mg
SDP
Catalog No. 502937095
Encompass_Preferred
An aglycone form of avermectin B1a; hyperpolarizes P. crassipes muscle fibers (MAC = 0.1 μM); decreases survival of C. elegans (MAC = 0.1 μM); prevents PTZ-induced seizures in mice (ED50 = 43.9 mg/kg)
Catalog No. 50-293-7095 Supplier Cayman Chemical Supplier No. 280511 MG
An aglycone form of avermectin B1a; hyperpolarizes P. crassipes muscle fibers (MAC = 0.1 μM); decreases survival of C. elegans (MAC = 0.1 μM); prevents PTZ-induced seizures in mice (ED50 = 43.9 mg/kg)