Learn More
Enterprise Technology Solutions Avery Direct Thermal Labels, 4"x3", 2000/BX, White
SDP
Catalog No. 5000740407
Encompass_Preferred
Universal Direct Thermal Labels work with your industrial printer to create and print one label or thousands at a time. They are excellent for high-volume printing. Use with Zebra, Intermec, Brady, Sato and Datamax industrial direct thermal printers. Permanent adhesive sticks and stays put. Create your labels with existing labeling software or Avery Design and Print.
Catalog No. 50-007-40407 Supplier Enterprise Technology Solutions Supplier No. AVE4134
Universal Direct Thermal Labels work with your industrial printer to create and print one label or thousands at a time. They are excellent for high-volume printing. Use with Zebra, Intermec, Brady, Sato and Datamax industrial direct thermal printers. Permanent adhesive sticks and stays put. Create your labels with existing labeling software or Avery Design and Print.