Learn More
Enterprise Technology Solutions Avery ID Labels, Self-Lam, 10UP, 1-1/32"x3-1/2", 250/PK, White
Supplier: Enterprise Technology Solutions AVE00757
Easy Align Self-Laminating ID Labels are tough enough to withstand harsh conditions. These labels come with a clear laminate that's easy to apply. Simply print the label and seal with the clear laminate film. With the Easy Align design, you can create perfect laminated labels without bubbles or wrinkles every time - no lamination machine required. UV-resistant material provides extra protection against exposure to sun and other harsh environments. They're also water-resistant so surfaces can be wiped clean. Labels resist scuffing, tearing and smudging so they remain intact and easy to read. The permanent adhesive sticks to a variety of surfaces, including plastic, wood, metal, paper, ceramic and glass. Use them to mark your property and equipment, identify contents of containers and organize shelving in warehouses, storage areas and work sites. Easily customize labels with free online templates and designs. Labels on letter-size sheets are compatible with inkjet and laser printers.