Learn More
Enterprise Technology Solutions Avery Labels, Removable, Surface Safe, 3-1/4"x8-3/8", 150/PK, WE
SDP
Supplier:  Enterprise Technology Solutions AVE61507
Encompass_Preferred
Rectangular labels feature a special adhesive that's engineered to stick strong and remove cleanly from smooth hard surfaces including metal, plastic and glass. Durable, professional-grade material resists water, chemicals and abrasion, and provides superior bar-code scanning. They are great for use in warehouses, offices, factories, hospitality and more. The removable adhesive sticks securely but removes cleanly for easy, on-demand changes. Labels are simple to customize with text, graphics or bar codes using the free online software from Avery Design & Print. They are recommended for indoor use. Labels are not recommended for use on walls. Sheets are compatible with both laser and inkjet printers. Each sheet includes three labels.
Catalog No. 50-008-32307