Learn More
Enterprise Technology Solutions Avery Labels,Round,SureFeed,Matte,PrinterSafe,1"Dia,1200/PK,White
SDP
Catalog No. 5000832624
Encompass_Preferred
Matte White 1" Round Labels offer the perfect blank canvas for your designs. Labels allow your colorful graphics and messaging to stand out with a polished and professional finish. Sure Feed technology reduces printer jams and smudging. Print-to-the-edge layout allows full-bleed designs all the way to the edge of labels. Easy Peel feature with Pop-Up Edges make label application quick and efficient. Use various online templates and design tools using Avery Design and Print Online. Customize your labels with Avery Design and Print Online and use various label templates and design tools to print custom labels on demand. Get crisp, smudge-free printing from both laser and inkjet printers. Create custom product labels, thank you favor labels, wax melt labels and more.
Catalog No. 50-008-32624 Supplier Enterprise Technology Solutions Supplier No. AVE05656
Matte White 1" Round Labels offer the perfect blank canvas for your designs. Labels allow your colorful graphics and messaging to stand out with a polished and professional finish. Sure Feed technology reduces printer jams and smudging. Print-to-the-edge layout allows full-bleed designs all the way to the edge of labels. Easy Peel feature with Pop-Up Edges make label application quick and efficient. Use various online templates and design tools using Avery Design and Print Online. Customize your labels with Avery Design and Print Online and use various label templates and design tools to print custom labels on demand. Get crisp, smudge-free printing from both laser and inkjet printers. Create custom product labels, thank you favor labels, wax melt labels and more.