Learn More
Grainger AVERY, Laser Label, 5267 25PK
Catalog No. NC2380749
Encompass_Preferred
The Avery Laser Label 5267 is a white, matte laser label sheet compatible with laser printers. It contains 30 labels per sheet in a letter size of 8.5 inches x 11 inches. Each label measures 1 inch x 2.63 inches. The adhesive is permanent pressure sensitive. The labels are made in the USA by Avery and are sold in packs of 100 sheets totaling 3000 labels. They work in laser printers and Avery states they have guaranteed smudge-proof printing. The labels have a temperature range of -20 to 160 degrees Fahrenheit. They can be used for general purpose labeling applications.
Catalog No. NC2380749 Supplier Grainger Supplier No. 5NHF8
The Avery Laser Label 5267 is a white, matte laser label sheet compatible with laser printers. It contains 30 labels per sheet in a letter size of 8.5 inches x 11 inches. Each label measures 1 inch x 2.63 inches. The adhesive is permanent pressure sensitive. The labels are made in the USA by Avery and are sold in packs of 100 sheets totaling 3000 labels. They work in laser printers and Avery states they have guaranteed smudge-proof printing. The labels have a temperature range of -20 to 160 degrees Fahrenheit. They can be used for general purpose labeling applications.