Learn More
Enterprise Technology Solutions Avery Mailing Labels, Weatherproof, 1-1/3"x4", 7000/BX, WE
SDP
Catalog No. 5000832681
Encompass_Preferred
Durable white polyester labels provide strong protection for harsh environments or conditions and resist moisture, scuffing, tearing and smudging. Ultrahold adhesive sticks to more tough surfaces ensuring your label doesn't fall off your package. Extra-strong adhesive adheres to metal, plastic, glass and more. Weatherproof labels with TrueBlock Technology are guaranteed to completely cover everything underneath so you can re-use mailing tubes, boxes and more. They are easy to customize with online free templates and designs using Avery Design and Print. The 8-1/2" x 11" sheets of labels are compatible with laser printers.
Catalog No. 50-008-32681 Supplier Enterprise Technology Solutions Supplier No. AVE95522
Durable white polyester labels provide strong protection for harsh environments or conditions and resist moisture, scuffing, tearing and smudging. Ultrahold adhesive sticks to more tough surfaces ensuring your label doesn't fall off your package. Extra-strong adhesive adheres to metal, plastic, glass and more. Weatherproof labels with TrueBlock Technology are guaranteed to completely cover everything underneath so you can re-use mailing tubes, boxes and more. They are easy to customize with online free templates and designs using Avery Design and Print. The 8-1/2" x 11" sheets of labels are compatible with laser printers.