Learn More
Enterprise Technology Solutions Avery Sign Labels,Perm,Laser,7"x10",15/PK,Yellow
SDP
Catalog No. 5000832660
Encompass_Preferred
Print incredibly durable, vinyl safety labels from any standard laser printer with these blank, self-adhesive, vinyl label sheets in vibrant, safety yellow. Ideal for outdoor and heavy-duty indoor applications, vinyl labels can be used for anything from warehouse labels to outdoor inspection labels and OSHA safety labels. They last up to 2 years outdoors and are waterproof, UV-resistant, fade-resistant and abrasion-resistant. Use for labeling applications in construction, warehouse, manufacturing and medical facilities. With strong permanent adhesive, they stick firmly to metal, plastic, glass and wood. The 3.4 mil vinyl film also conforms to rough and curved surfaces like concrete, cinderblock and industrial tubing. Avery templates for power source labels, biohazard signs, Prop 65 warnings and OSHA safety sign labels are easy to customize using Avery's cloud-based, industrial design software. Each sheet includes one label (7" x 10").
Catalog No. 50-008-32660 Supplier Enterprise Technology Solutions Supplier No. AVE61554
Print incredibly durable, vinyl safety labels from any standard laser printer with these blank, self-adhesive, vinyl label sheets in vibrant, safety yellow. Ideal for outdoor and heavy-duty indoor applications, vinyl labels can be used for anything from warehouse labels to outdoor inspection labels and OSHA safety labels. They last up to 2 years outdoors and are waterproof, UV-resistant, fade-resistant and abrasion-resistant. Use for labeling applications in construction, warehouse, manufacturing and medical facilities. With strong permanent adhesive, they stick firmly to metal, plastic, glass and wood. The 3.4 mil vinyl film also conforms to rough and curved surfaces like concrete, cinderblock and industrial tubing. Avery templates for power source labels, biohazard signs, Prop 65 warnings and OSHA safety sign labels are easy to customize using Avery's cloud-based, industrial design software. Each sheet includes one label (7" x 10").