Learn More
Medchemexpress LLC HY-101864 5mg Medchemexpress, AVex-73 hydrochloride CAS:195615-84-0 Purity:>98%
SDP
Catalog No. 501967898
Encompass_Preferred
Medchemexpress, HY-101864 5mg AVex-73 HCl CAS:195615-84-0 AVex-73 hydrochloride is a Sigma-1 Receptor agonist with an IC50 of 860 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7898 Supplier Medchemexpress LLC Supplier No. HY1018645MG
Medchemexpress, HY-101864 5mg AVex-73 HCl CAS:195615-84-0 AVex-73 hydrochloride is a Sigma-1 Receptor agonist with an IC50 of 860 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.