Learn More
Selleck Chemical LLC Avexitide-1mg
SDP
Supplier:  Selleck Chemical LLC P12031MG
Encompass_Preferred
Avexitide (Exendin (9-39)) is a specific and competitive antagonist of glucagon-like peptide-1 (GLP-1) receptor.
Catalog No. 50-238-9612