Learn More
TARGETMOL CHEMICALS INC AVEXITIDE 5MG
Supplier: TARGETMOL CHEMICALS INC TP21005MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Avexitide (Exendin-3 (9-39) amide) (Exendin (9-39)) is a specific and competitive antagonist of glucagon-like peptide-1 (GLP-1) receptor. purity: 98%