Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent Technologies AVF 60 Gold Vacuum Pump Oil, 1 L

Catalog No. NC2948553
Encompass

AVF 60 Gold is a high-quality liquid reagent formulated specifically for use in analytical chemistry applications, including as a vacuum pump oil. It is designed for compatibility with LC/MS vacuum pumps, such as the Agilent MS40+ and Edwards E2Mxx series, where it supports consistent performance by maintaining optimal rough vacuum conditions necessary for reliable ion transmission.

  • Supports vacuum pump performance in analytical systems
  • Optimized for use with specific LC/MS vacuum pump models
  • Liquid formulation with a pH of 8 to 9
  • Soluble in both cold and hot water
  • Does not contain substances classified as PBT or vPvB
  • Contains no SVHC compounds

Catalog No. NC2948553 Supplier Agilent Technologies Supplier No. 60401444
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AVF 60 Gold is a high-quality liquid reagent formulated specifically for use in analytical chemistry applications, including as a vacuum pump oil. It is designed for compatibility with LC/MS vacuum pumps, such as the Agilent MS40+ and Edwards E2Mxx series, where it supports consistent performance by maintaining optimal rough vacuum conditions necessary for reliable ion transmission.

  • Supports vacuum pump performance in analytical systems
  • Optimized for use with specific LC/MS vacuum pump models
  • Liquid formulation with a pH of 8 to 9
  • Soluble in both cold and hot water
  • Does not contain substances classified as PBT or vPvB
  • Contains no SVHC compounds

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.