Learn More
TARGETMOL CHEMICALS INC AVG-233 1MG
SDP
Catalog No. 502877064
Encompass_Preferred
AVG-233 is an orally available RNA-dependent RNA polymerase (RdRp) inhibitor with antiviral activity that blocks respiratory syncytial virus and SARS-CoV-2 replication.AVG-233 is used in the study of respiratory syncytial virus infections. Purity 99%
Catalog No. 50-287-7064 Supplier TARGETMOL CHEMICALS INC Supplier No. T632531MG
AVG-233 is an orally available RNA-dependent RNA polymerase (RdRp) inhibitor with antiviral activity that blocks respiratory syncytial virus and SARS-CoV-2 replication.AVG-233 is used in the study of respiratory syncytial virus infections. Purity 99%