Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Avg-233 | 2151937-80-1 | 98.1% | 100 MG
SDP

Catalog No. 5000232178
Encompass_Preferred

AVG-233 is a potent, orally active RNA-dependent RNA polymerase (RdRp) inhibitor used for research into respiratory syncytial virus (RSV). It prevents initiation of the viral polymerase complex and shows nanomolar activity against RSV strains and clinical isolates.

  • Potent RNA-dependent RNA polymerase inhibitor.
  • Reported EC50 = 0.14-0.31 μM against RSV strains and clinical isolates.
  • Molecular formula: C26H22ClN5O3.
  • Molecular weight: 487.94 g/mol.
  • Available as a solid in multiple pack sizes (5 mg-500 mg).
  • Intended for research use only; not for human or diagnostic use.

Catalog No. 50-002-32178 Supplier Medchemexpress LLC Supplier No. HY122587100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

AVG-233 is a potent, orally active RNA-dependent RNA polymerase (RdRp) inhibitor used for research into respiratory syncytial virus (RSV). It prevents initiation of the viral polymerase complex and shows nanomolar activity against RSV strains and clinical isolates.

  • Potent RNA-dependent RNA polymerase inhibitor.
  • Reported EC50 = 0.14-0.31 μM against RSV strains and clinical isolates.
  • Molecular formula: C26H22ClN5O3.
  • Molecular weight: 487.94 g/mol.
  • Available as a solid in multiple pack sizes (5 mg-500 mg).
  • Intended for research use only; not for human or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.