Learn More
Medchemexpress LLC Avg-233 | 2151937-80-1 | 98.1% | 25 MG
SDP
Catalog No. 5000232181
Encompass_Preferred
AVG-233 (CAS 2151937-80-1) is a small-molecule RNA-dependent RNA polymerase (RdRp) inhibitor used for research on respiratory syncytial virus (RSV). It prevents initiation of the viral polymerase complex and shows nanomolar activity against RSV isolates (reported EC50 = 0.14-0.31 μM). Supplied as an off-white to yellow solid for laboratory use.
  • Potent RdRp inhibitory activity (RSV EC50 = 0.14-0.31 μM).
  • High purity solid, typically reported around 98%.
  • Molecular formula C26H22ClN5O3; molecular weight 487.94.
  • Available in multiple small-scale pack sizes suitable for research.
  • Intended for in vitro and preclinical research; follow safety and storage recommendations.
Catalog No. 50-002-32181 Supplier Medchemexpress LLC Supplier No. HY12258725MG
AVG-233 (CAS 2151937-80-1) is a small-molecule RNA-dependent RNA polymerase (RdRp) inhibitor used for research on respiratory syncytial virus (RSV). It prevents initiation of the viral polymerase complex and shows nanomolar activity against RSV isolates (reported EC50 = 0.14-0.31 μM). Supplied as an off-white to yellow solid for laboratory use.
  • Potent RdRp inhibitory activity (RSV EC50 = 0.14-0.31 μM).
  • High purity solid, typically reported around 98%.
  • Molecular formula C26H22ClN5O3; molecular weight 487.94.
  • Available in multiple small-scale pack sizes suitable for research.
  • Intended for in vitro and preclinical research; follow safety and storage recommendations.