Learn More
Medchemexpress LLC Avg-233 | 2151937-80-1 | 98.1% | 5 MG
SDP
Catalog No. 5000230807
Encompass_Preferred
AVG-233 is a potent, orally active RNA-dependent RNA polymerase (RdRp) inhibitor used for research on respiratory syncytial virus (RSV). It prevents initiation of the viral polymerase complex and shows nanomolar antiviral activity against RSV strains and clinical isolates (EC50 = 0.14-0.31 μM).
  • Potent RdRp inhibition with nanomolar activity (EC50 = 0.14-0.31 μM).
  • Orally active small molecule appropriate for in vitro and in vivo RSV research.
  • Solid, off-white to yellow powder; store at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).
  • High reported purity from suppliers (≈98-99%).
  • Available in small milligram quantities and as a DMSO solution for screening.
Catalog No. 50-002-30807 Supplier Medchemexpress LLC Supplier No. HY1225875MG
AVG-233 is a potent, orally active RNA-dependent RNA polymerase (RdRp) inhibitor used for research on respiratory syncytial virus (RSV). It prevents initiation of the viral polymerase complex and shows nanomolar antiviral activity against RSV strains and clinical isolates (EC50 = 0.14-0.31 μM).
  • Potent RdRp inhibition with nanomolar activity (EC50 = 0.14-0.31 μM).
  • Orally active small molecule appropriate for in vitro and in vivo RSV research.
  • Solid, off-white to yellow powder; store at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).
  • High reported purity from suppliers (≈98-99%).
  • Available in small milligram quantities and as a DMSO solution for screening.