Learn More
Medchemexpress LLC Avg-233 | 2151937-80-1 | 98.9% | 50 MG
SDP
Catalog No. 5000232028
Encompass_Preferred
AVG-233 is a small-molecule RNA-dependent RNA polymerase (RdRp) inhibitor used for research on respiratory syncytial virus (RSV). It demonstrates nanomolar antiviral potency and is supplied for laboratory research applications.
  • Potent RdRp inhibitory activity against RSV (EC50 0.14-0.31 μM).
  • High chemical purity suitable for research (reported ~98.9%).
  • Molecular formula C26H22ClN5O3; molecular weight 487.94.
  • Supplied as a 50 mg vial for laboratory use.
  • Intended for in vitro research applications.
Catalog No. 50-002-32028 Supplier Medchemexpress LLC Supplier No. HY12258750MG
AVG-233 is a small-molecule RNA-dependent RNA polymerase (RdRp) inhibitor used for research on respiratory syncytial virus (RSV). It demonstrates nanomolar antiviral potency and is supplied for laboratory research applications.
  • Potent RdRp inhibitory activity against RSV (EC50 0.14-0.31 μM).
  • High chemical purity suitable for research (reported ~98.9%).
  • Molecular formula C26H22ClN5O3; molecular weight 487.94.
  • Supplied as a 50 mg vial for laboratory use.
  • Intended for in vitro research applications.