Learn More
AdipoGen AVG-Cl
SDP
Supplier:  AdipoGen CDXA0185M025
Encompass_Preferred
Chemical. CAS 55720-26-8. Formula C6H12N2O3 . HCl. MW 196.63. Synthetic. Potent inhibitor of ethylene synthesis in plants acting at the level of 1-aminocyclopropanecarboxylic acid synthase.
Catalog No. 50-256-1807