Learn More
Novus Biologicals™ Avian Influenza A H7N9 Hemagglutinin Antibody Pair [HRP]

Description
Specifications
Specifications
| Assay Range | 62.5 pg/mL |
| Assay Sensitivity | 62.5-4000 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes an antibody specific for Influenza A H7N9 (A/Anhui/1/2013) Hemagglutinin. |
| Target Species | Virus |
| For Use With (Application) | Sandwich ELISA |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.