Learn More
ATCC Avian leukosis virus; Strain: RAV-2; Rous-associated virus-2
Catalog No. 502384172
Encompass
Avian leukosis virus strain RAV-2 is propagated in UMNSAH/DF-1 cells (ATCC CRL-12203).
Catalog No. 50-238-4172 Supplier ATCC Supplier No. VR1828
Avian leukosis virus strain RAV-2 is propagated in UMNSAH/DF-1 cells (ATCC CRL-12203).