Learn More
Med Vet International Avian Oral Speculum, J0051S, Set of 3
Catalog No. 5000584488
Encompass_Preferred
The safe and effective way to examine the avian oral cavity; also aids in the administration of medication using the JOR194 series JorVet feeding needles. Stainless steel.
Catalog No. 50-005-84488 Supplier Med Vet International Supplier No. JOR51S
The safe and effective way to examine the avian oral cavity; also aids in the administration of medication using the JOR194 series JorVet feeding needles. Stainless steel.