Learn More
Med Vet International Avian Oral Speculum, large, J0051Z
Catalog No. 5000585065
Encompass_Preferred
The safe and effective way to examine the avian oral cavity; also aids in the administration of medication using the JOR194 series JorVet feeding needles. Stainless steel.
Catalog No. 50-005-85065 Supplier Med Vet International Supplier No. JOR51Z
The safe and effective way to examine the avian oral cavity; also aids in the administration of medication using the JOR194 series JorVet feeding needles. Stainless steel.