Learn More
Med Vet International Avian Oral Speculum, Small. 4 3/4" x 1/4". (i.e., parakeets), J0051X
Catalog No. 5000585085
Encompass_Preferred
The safe and effective way to examine the avian oral cavity; also aids in the administration of medication using the JOR194 series JorVet feeding needles. Stainless steel.
Catalog No. 50-005-85085 Supplier Med Vet International Supplier No. JOR51X
The safe and effective way to examine the avian oral cavity; also aids in the administration of medication using the JOR194 series JorVet feeding needles. Stainless steel.