Learn More
Med Vet International Avian/Small Exotic Cole Endotracheal Tube 2.5mm, J0152B
Catalog No. 5000583020
Encompass_Preferred
Perfect for the smallest of patients. Lower profile on the distal end adds proper placement, and larger proximal end helps form a tighter seal. Steppeddown "Cole"-style wall. Silicon. Complete with attached standard 15mm connector.
Catalog No. 50-005-83020 Supplier Med Vet International Supplier No. JOR152B
Perfect for the smallest of patients. Lower profile on the distal end adds proper placement, and larger proximal end helps form a tighter seal. Steppeddown "Cole"-style wall. Silicon. Complete with attached standard 15mm connector.