Learn More
MSC CRC Aviation T-Force Degreaser, Aerosol, 18 oz Net Fill, Non-Flammable
Catalog No. 502837456
Encompass
This aviation degreaser is a heavy-duty, non-flammable cleaner that is designed to evaporate quickly and leave no residue. It is airframe approved and functions as a non-HAP degreaser, making it suitable for a variety of precision cleaning applications.
  • Evaporates quickly and leaves no residue
  • Airframe approved
  • Non-HAP degreaser
  • Has no flash or fire point
Catalog No. 50-283-7456 Supplier MSC Supplier No. 36544054
This aviation degreaser is a heavy-duty, non-flammable cleaner that is designed to evaporate quickly and leave no residue. It is airframe approved and functions as a non-HAP degreaser, making it suitable for a variety of precision cleaning applications.
  • Evaporates quickly and leaves no residue
  • Airframe approved
  • Non-HAP degreaser
  • Has no flash or fire point