Learn More
Checkers Aviation Wheel Chocks for Small/Mid-Sized Aircraft AC3500 Checkers
Shop All Justrite Safety Group Products

Description
- Checkers™ Aviation Wheel Chocks include the AC series, made of highly durable, lightweight polyurethane
- AC Chocks last longer than wood chocks, never need painting and eliminate Foreign Object Damage threats for aircraft
- The AC Series is ideal for private, commercial, cargo and military aircraft
Specifications
Specifications
| Dimensions (L x W x H) | 12 x 5 x 6 in. |
| For Use With (Equipment) | Small/Mid-Sized Aircraft |
| Color | Yellow |
| Material | Polyurethane |
| Product Type | Wheel Chock |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.