Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Checkers Aviation Wheel Chocks
GREENER_CHOICE

Catalog No. 19088482 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Lime Green
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-088-482 Lime Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-088-482 Supplier Checkers Supplier No. AC4614LRCHFLG
Add to Cart
Add to Cart

Aviation Wheel Chocks include the AC series, made of highly durable, lightweight polyurethane.

  • Checkers™ Aviation Wheel Chocks include the AC series, made of highly durable, lightweight polyurethane
  • AC Chocks last longer than wood chocks, never need painting and eliminate Foreign Object Damage threats for aircraft
  • The AC Series is ideal for private, commercial, cargo and military aircraft

Specifications

Product Type Wheel Chock
Dimensions (L x W x H) 14.5 x 6 x 4 in.
Material Urethane
For Use With (Equipment) Private, Commercial, Cargo, and Military Aircrafts
Color Lime Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.