Learn More
Medchemexpress LLC Avibactam free acid | 1192500-31-4 | 99.27% | 265.24 | 50 MG
SDP
Catalog No. 5000359892
Encompass_Preferred
Avibactam free acid is a covalent and reversible non-β-lactam β-lactamase inhibitor. It effectively inhibits β-lactamase TEM-1 and CTX-M-15.
  • Inhibits class A and C β-lactamases
  • Has little antibacterial activity
  • Does not inhibit metallo-type β-lactamases or Acinetobacter OXA carbapenemases
Catalog No. 50-003-59892 Supplier Medchemexpress LLC Supplier No. HY1487950MG
Add to Cart
Add to Cart
Avibactam free acid is a covalent and reversible non-β-lactam β-lactamase inhibitor. It effectively inhibits β-lactamase TEM-1 and CTX-M-15.
  • Inhibits class A and C β-lactamases
  • Has little antibacterial activity
  • Does not inhibit metallo-type β-lactamases or Acinetobacter OXA carbapenemases