Learn More
Medchemexpress LLC Avibactam free acid | 1192500-31-4 | 99.3% | 265.24 | 100 MG
SDP
Catalog No. 5000361672
Encompass_Preferred
Avibactam free acid is a covalent and reversible non-β-lactam β-lactamase inhibitor that inhibits β-lactamase TEM-1 and CTX-M-15.
  • Covalent and reversible non-β-lactam β-lactamase inhibitor.
  • Inhibits β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively.
  • Exhibits little antibacterial activity.
  • Inhibits class A and C β-lactamases, but not metallo types and Acinetobacter OXA carbapenemases.
Catalog No. 50-003-61672 Supplier Medchemexpress LLC Supplier No. HY14879100MG
Add to Cart
Add to Cart
Avibactam free acid is a covalent and reversible non-β-lactam β-lactamase inhibitor that inhibits β-lactamase TEM-1 and CTX-M-15.
  • Covalent and reversible non-β-lactam β-lactamase inhibitor.
  • Inhibits β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively.
  • Exhibits little antibacterial activity.
  • Inhibits class A and C β-lactamases, but not metallo types and Acinetobacter OXA carbapenemases.