Learn More
Medchemexpress LLC Avibactam free acid | 1192500-31-4 | 99.3% | 265.24 | 5 MG
SDP
Catalog No. 5000359727
Encompass_Preferred
Avibactam free acid is a covalent and reversible non-β-lactam β-lactamase inhibitor. It effectively inhibits β-lactamase TEM-1 and CTX-M-15, with IC50s of 8 nM and 5 nM respectively. This product exhibits minimal antibacterial activity but targets class A and C β-lactamases, excluding metallo types and Acinetobacter OXA carbapenemases. It is intended for research use only.
  • Covalent and reversible non-β-lactam β-lactamase inhibitor
  • Inhibits β-lactamase TEM-1 and CTX-M-15
  • Targets class A and C β-lactamases
  • Minimal antibacterial activity
  • Suitable for research applications
Catalog No. 50-003-59727 Supplier Medchemexpress LLC Supplier No. HY148795MG
Avibactam free acid is a covalent and reversible non-β-lactam β-lactamase inhibitor. It effectively inhibits β-lactamase TEM-1 and CTX-M-15, with IC50s of 8 nM and 5 nM respectively. This product exhibits minimal antibacterial activity but targets class A and C β-lactamases, excluding metallo types and Acinetobacter OXA carbapenemases. It is intended for research use only.
  • Covalent and reversible non-β-lactam β-lactamase inhibitor
  • Inhibits β-lactamase TEM-1 and CTX-M-15
  • Targets class A and C β-lactamases
  • Minimal antibacterial activity
  • Suitable for research applications