Learn More
Medchemexpress LLC Avibactam (NXL-104) free acid | 1192500-31-4 | MFCD30478396 | 99.3% | 265.24 g/mol | C7H11N3O6S | 10 MG
SDP
Catalog No. 5000285343
Encompass_Preferred
Avibactam free acid is a covalent, reversible non-β-lactam β-lactamase inhibitor used as a research reagent. It inhibits class A and C β-lactamases, including TEM-1 and CTX-M-15 (reported IC50s 8 nM and 5 nM), shows little intrinsic antibacterial activity, and does not inhibit metallo-β-lactamases or certain OXA carbapenemases. The material is supplied as a white to off-white solid with high purity and defined storage conditions.
  • Inhibits TEM-1 and CTX-M-15 with low-nanomolar IC50s.
  • Covalent and reversible mechanism of inhibition.
  • Active against class A and C β-lactamases.
  • Not active against metallo-β-lactamases or OXA carbapenemases.
  • High purity suitable for biochemical and enzymatic assays.
  • Stable powder with recommended cold storage for long-term retention.
Catalog No. 50-002-85343 Supplier Medchemexpress LLC Supplier No. HY1487910MG
Avibactam free acid is a covalent, reversible non-β-lactam β-lactamase inhibitor used as a research reagent. It inhibits class A and C β-lactamases, including TEM-1 and CTX-M-15 (reported IC50s 8 nM and 5 nM), shows little intrinsic antibacterial activity, and does not inhibit metallo-β-lactamases or certain OXA carbapenemases. The material is supplied as a white to off-white solid with high purity and defined storage conditions.
  • Inhibits TEM-1 and CTX-M-15 with low-nanomolar IC50s.
  • Covalent and reversible mechanism of inhibition.
  • Active against class A and C β-lactamases.
  • Not active against metallo-β-lactamases or OXA carbapenemases.
  • High purity suitable for biochemical and enzymatic assays.
  • Stable powder with recommended cold storage for long-term retention.