Learn More
Medchemexpress LLC HY-14879A 200mg , Avibactam (sodium) NXL-104 CAS:1192491-61-4 Purity:98%
SDP
Catalog No. 501492532
Encompass_Preferred
Medchemexpress, HY-14879A 200mg Avibactam (sodium) NXL-104 CAS:1192491-61-4 Formula:C7H10N3NaO6S IC50: 8 nM (TEM-1), 5 nM (CTX-M-15) Purity:98% Avibactam sodium is a covalent, reversible β-lactamase inhibitor, inhibits β-lactamase TEM-1 and CTX-M-15 with IC 50 of 8 nM and 5 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2532 Supplier Medchemexpress LLC Supplier No. HY14879A200MG
Medchemexpress, HY-14879A 200mg Avibactam (sodium) NXL-104 CAS:1192491-61-4 Formula:C7H10N3NaO6S IC50: 8 nM (TEM-1), 5 nM (CTX-M-15) Purity:98% Avibactam sodium is a covalent, reversible β-lactamase inhibitor, inhibits β-lactamase TEM-1 and CTX-M-15 with IC 50 of 8 nM and 5 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.