Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Avibactam sodium | 1192491-61-4 | 99.9% | 287.23 | 25 MG
SDP

Catalog No. 5000363542
Encompass_Preferred

Avibactam sodium is the analytical standard of Avibactam sodium. It is a covalent and reversible non-β-lactam β-lactamase inhibitor, inhibiting β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively. This product is intended for research and analytical applications, and is for research use only.

  • Purity: 99.86%
  • Application: analytical standard, reference standard supplied assay
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
  • Molecular weight: 287.23
  • Formula: C7H10N3NaO6S
  • Appearance: solid
  • Color: white to off-white

Catalog No. 50-003-63542 Supplier Medchemexpress LLC Supplier No. 25MGHY14879AR
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Avibactam sodium is the analytical standard of Avibactam sodium. It is a covalent and reversible non-β-lactam β-lactamase inhibitor, inhibiting β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively. This product is intended for research and analytical applications, and is for research use only.

  • Purity: 99.86%
  • Application: analytical standard, reference standard supplied assay
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
  • Molecular weight: 287.23
  • Formula: C7H10N3NaO6S
  • Appearance: solid
  • Color: white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.