Learn More
Medchemexpress LLC Avibactam sodium | 1192491-61-4 | 99.99% | 287.23 | 1 ML
SDP
Catalog No. 5000361554
Encompass_Preferred
Avibactam sodium (NXL-104) is a covalent and reversible non-β-lactam β-lactamase inhibitor. It inhibits β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively. This molecule exhibits little antibacterial activity and effectively inhibits class A and C β-lactamases, but not metallo types and Acinetobacter OXA carbapenemases. It is intended for research use only.
  • Covalent and reversible non-β-lactam β-lactamase inhibitor
  • Inhibits β-lactamase TEM-1 and CTX-M-15
  • Exhibits little antibacterial activity
  • Inhibits class A and C β-lactamases
  • Not effective against metallo types and Acinetobacter OXA carbapenemases
  • Intended for research use only
Catalog No. 50-003-61554 Supplier Medchemexpress LLC Supplier No. HY14879A10MM/1ML
Avibactam sodium (NXL-104) is a covalent and reversible non-β-lactam β-lactamase inhibitor. It inhibits β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively. This molecule exhibits little antibacterial activity and effectively inhibits class A and C β-lactamases, but not metallo types and Acinetobacter OXA carbapenemases. It is intended for research use only.
  • Covalent and reversible non-β-lactam β-lactamase inhibitor
  • Inhibits β-lactamase TEM-1 and CTX-M-15
  • Exhibits little antibacterial activity
  • Inhibits class A and C β-lactamases
  • Not effective against metallo types and Acinetobacter OXA carbapenemases
  • Intended for research use only