Learn More
Selleck Chemical LLC Avibactam sodium S3732-100mg
SDP
Catalog No. 503109185
Encompass_Preferred
Avibactam (AVE-1330A NXL104) is a covalent reversible non- -lactam -lactamase inhibitor with IC50 values of 8 80 and 38 nM for TEM-1 P99 and KPC-2 -lactamases respectively
Catalog No. 50-310-9185 Supplier Selleck Chemical LLC Supplier No. S3732100MG
Avibactam (AVE-1330A NXL104) is a covalent reversible non- -lactam -lactamase inhibitor with IC50 values of 8 80 and 38 nM for TEM-1 P99 and KPC-2 -lactamases respectively