Learn More
TARGETMOL CHEMICALS INC AVIBACTAM SODIUM 50MG
SDP
Catalog No. 502576791
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Avibactam sodium (NXL-104) is a reversible (beta)-lactamase inhibitor (IC50 8/80/38 nM for TEM-1/P99/KPC-2 (beta)-lactamases). purity: 99%
Catalog No. 50-257-6791 Supplier TARGETMOL CHEMICALS INC Supplier No. T420150MG
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Avibactam sodium (NXL-104) is a reversible (beta)-lactamase inhibitor (IC50 8/80/38 nM for TEM-1/P99/KPC-2 (beta)-lactamases). purity: 99%