Learn More
Medchemexpress LLC HY-14879A 50mg , Avibactam (sodium) CAS:1192491-61-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14879A50MG
Encompass_Preferred
HY-14879A 50mg Avibactam (sodium) CAS: 1192491-61-4 Avibactam sodium (NXL-104) is a covalent and reversible non-β-lactam β-lactamase inhibitor which inhibits β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9362