Learn More
Medchemexpress LLC Avibactam sodium hydrate | 2938989-90-1 | MFCD30533174 | 99.6% | 305.24 | 100 MG
SDP
Catalog No. 5000359857
Encompass_Preferred
Avibactam sodium hydrate is a covalent and reversible non-β-lactam β-lactamase inhibitor which inhibits β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively. It is for research use only.
  • Covalent and reversible non-β-lactam β-lactamase inhibitor
  • Inhibits β-lactamase TEM-1 and CTX-M-15
  • Active against class A and C β-lactamases
  • Shows significant effect on bacteria in *K. pneumoniae* infected mouse models
Catalog No. 50-003-59857 Supplier Medchemexpress LLC Supplier No. HY14879B100MG
Avibactam sodium hydrate is a covalent and reversible non-β-lactam β-lactamase inhibitor which inhibits β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively. It is for research use only.
  • Covalent and reversible non-β-lactam β-lactamase inhibitor
  • Inhibits β-lactamase TEM-1 and CTX-M-15
  • Active against class A and C β-lactamases
  • Shows significant effect on bacteria in *K. pneumoniae* infected mouse models