Learn More
Medchemexpress LLC Avibactam sodium hydrate (NXL-104 hydrate) | 2938989-90-1 | 99.6% | 305.24 | 50 MG
SDP
Catalog No. 5000361576
Encompass_Preferred
Avibactam sodium hydrate (NXL-104) is a covalent and reversible non-β-lactam β-lactamase inhibitor which inhibits β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively. It is intended for research use only.
  • Covalent and reversible non-β-lactam β-lactamase inhibitor
  • Inhibits β-lactamase TEM-1 and CTX-M-15
  • IC50 of 5 nM for CTX-M-15 and 8 nM for TEM-1
  • For research use only
Catalog No. 50-003-61576 Supplier Medchemexpress LLC Supplier No. HY14879B50MG
Add to Cart
Add to Cart
Avibactam sodium hydrate (NXL-104) is a covalent and reversible non-β-lactam β-lactamase inhibitor which inhibits β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively. It is intended for research use only.
  • Covalent and reversible non-β-lactam β-lactamase inhibitor
  • Inhibits β-lactamase TEM-1 and CTX-M-15
  • IC50 of 5 nM for CTX-M-15 and 8 nM for TEM-1
  • For research use only