Learn More
Medchemexpress LLC Avibactam sodium salt-13C5 | 1192491-61-4 | 99.9% | 292.19 | 1 MG
SDP
Catalog No. 5000359725
Encompass_Preferred
Avibactam sodium salt-13C5 (NXL-104-13C5) is a 13C labeled version of Avibactam sodium (NXL-104). It is a covalent and reversible non-β-lactam β-lactamase inhibitor with inhibitory activity against β-lactamase TEM-1 and CTX-M-15 (IC50 values of 8 nM and 5 nM, respectively). This compound is intended for research use only and serves as a tracer or an internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS. Stable heavy isotopes, such as carbon-13, are incorporated into drug molecules primarily for quantitation during drug development.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Exhibits covalent and reversible non-β-lactam β-lactamase inhibition
  • Inhibits β-lactamase TEM-1 and CTX-M-15
  • For research use only
Catalog No. 50-003-59725 Supplier Medchemexpress LLC Supplier No. HY14879S21MG
Avibactam sodium salt-13C5 (NXL-104-13C5) is a 13C labeled version of Avibactam sodium (NXL-104). It is a covalent and reversible non-β-lactam β-lactamase inhibitor with inhibitory activity against β-lactamase TEM-1 and CTX-M-15 (IC50 values of 8 nM and 5 nM, respectively). This compound is intended for research use only and serves as a tracer or an internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS. Stable heavy isotopes, such as carbon-13, are incorporated into drug molecules primarily for quantitation during drug development.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Exhibits covalent and reversible non-β-lactam β-lactamase inhibition
  • Inhibits β-lactamase TEM-1 and CTX-M-15
  • For research use only