Learn More
Medchemexpress LLC Avibactam sodium (Standard) | 1192491-61-4 | 99.86% | 287.23 | 100 MG
SDP
Catalog No. 5000360812
Encompass_Preferred
Avibactam (sodium) (Standard) is the analytical standard of Avibactam (sodium) intended for research and analytical applications. It is a covalent and reversible non-β-lactam β-lactamase inhibitor which inhibits β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively. The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • Commonly used in qualitative, quantitative and methodological research experiments
  • Reference standard for assay
  • Utilized in HPLC, GC, and MS experiments
Catalog No. 50-003-60812 Supplier Medchemexpress LLC Supplier No. HY14879AR100MG
Avibactam (sodium) (Standard) is the analytical standard of Avibactam (sodium) intended for research and analytical applications. It is a covalent and reversible non-β-lactam β-lactamase inhibitor which inhibits β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively. The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • Commonly used in qualitative, quantitative and methodological research experiments
  • Reference standard for assay
  • Utilized in HPLC, GC, and MS experiments