Learn More
Medchemexpress LLC Avibactam sodium (Standard) | 1192491-61-4 | 99.9% | 287.23 | 5 MG
SDP
Catalog No. 5000359890
Encompass_Preferred
Avibactam (sodium) (Standard) is the analytical standard of Avibactam (sodium), intended for research and analytical applications. Avibactam sodium (NXL-104) is described as a covalent and reversible non-β-lactam β-lactamase inhibitor. It inhibits β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively.
  • Analytical standard for Avibactam (sodium).
  • Intended for research and analytical applications.
  • Covalent and reversible non-β-lactam β-lactamase inhibitor.
  • Inhibits β-lactamase TEM-1 and CTX-M-15.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-003-59890 Supplier Medchemexpress LLC Supplier No. HY14879AR5MG
Avibactam (sodium) (Standard) is the analytical standard of Avibactam (sodium), intended for research and analytical applications. Avibactam sodium (NXL-104) is described as a covalent and reversible non-β-lactam β-lactamase inhibitor. It inhibits β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively.
  • Analytical standard for Avibactam (sodium).
  • Intended for research and analytical applications.
  • Covalent and reversible non-β-lactam β-lactamase inhibitor.
  • Inhibits β-lactamase TEM-1 and CTX-M-15.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.