Learn More
Medchemexpress LLC Avibactam tomilopil | 2245880-46-8 | 98.6% | 393.41 | 5 MG
SDP
Catalog No. 5000364349
Encompass_Preferred
Avibactam tomilopil is an orally active prodrug of Avibactam and a β-lactamase inhibitor. It can be used in the study of diseases such as urinary tract infections.
  • Acts as an orally active prodrug of Avibactam
  • Functions as a β-lactamase inhibitor
  • Useful for studying diseases like urinary tract infections
  • Converts into active Avibactam in vivo when combined with Ceftibuten for oral dosing
  • Avibactam blocks enzymes produced by gram-negative bacteria
  • Protects β-lactam antibiotics from degradation
Catalog No. 50-003-64349 Supplier Medchemexpress LLC Supplier No. 5MGHY132987
Avibactam tomilopil is an orally active prodrug of Avibactam and a β-lactamase inhibitor. It can be used in the study of diseases such as urinary tract infections.
  • Acts as an orally active prodrug of Avibactam
  • Functions as a β-lactamase inhibitor
  • Useful for studying diseases like urinary tract infections
  • Converts into active Avibactam in vivo when combined with Ceftibuten for oral dosing
  • Avibactam blocks enzymes produced by gram-negative bacteria
  • Protects β-lactam antibiotics from degradation