Learn More
Miles Scientific Avicel F | 250um | 20X20cm | Scored | 25/bx
SDP
Catalog No. NC1806904
Encompass_Preferred
These plates are coated with AVICEL microcrystalline cellulose manufactured by the American Viscose division of FMC Corp. The adsorbent consists of regenerated alpha cellulose particles of nearly spherical shape and 50 micron mean diameter. 
Catalog No. NC1806904 Supplier Miles Scientific Supplier No. P06511
These plates are coated with AVICEL microcrystalline cellulose manufactured by the American Viscose division of FMC Corp. The adsorbent consists of regenerated alpha cellulose particles of nearly spherical shape and 50 micron mean diameter. 