Learn More
Medchemexpress LLC HY-N0222 5mg Medchemexpress, Avicularin CAS:572-30-5 Purity:>98%
SDP
Catalog No. 502031316
Encompass_Preferred
Medchemexpress, HY-N0222 5mg Avicularin CAS:572-30-5 Avicularin is a bio-active flavonoid from plants, anti-inflammatory, anti-allergic, anti-oxidant, hepatoprotective, and anti-tumor activities. Avicularin exhibits anti-inflammatory activity through the suppression of ERK signaling pathway in LPS-stimulated RAW 264.7 macrophage cells. Avicularin ameliorates human hepatocellular carcinoma via the regulation of NF κB (p65), COX 2 and PPAR γ activities[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1316 Supplier Medchemexpress LLC Supplier No. HYN02225MG
Medchemexpress, HY-N0222 5mg Avicularin CAS:572-30-5 Avicularin is a bio-active flavonoid from plants, anti-inflammatory, anti-allergic, anti-oxidant, hepatoprotective, and anti-tumor activities. Avicularin exhibits anti-inflammatory activity through the suppression of ERK signaling pathway in LPS-stimulated RAW 264.7 macrophage cells. Avicularin ameliorates human hepatocellular carcinoma via the regulation of NF κB (p65), COX 2 and PPAR γ activities[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.